missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific E-Z LOAD™ Histosette™ II Biopsy Cassette Lids

Catalog No. 0550121
Change view
Click to view available options
Color:
Lilac
Orange
Tan
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
0550121 Tan
0550123 Lilac
0550124 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 0550121 Supplier Simport™ Scientific Supplier No. M4838LI

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

E-Z LOAD™ Histosette™ II Biopsy Cassette Lids for use with Micro Writer™, Lamb™ and SHUR/Mark™ cassette labeling instruments.

  • For use with Micro Writer™, Lamb™ and SHUR/Mark™ cassette labeling instruments
  • Stacks of 50 cassettes each; case contains two boxes of 20 stacks
  • Bases and covers must be ordered separately

Specifications

Color Tan
Product Type Biopsy Cassette Cover