Learn More
Simport™ Scientific CoreDish™ Biopsy Containers
CoreDish™ Biopsy Containers allow for safe and easy transport of specimens from collection to analysis.
Supplier: Simport™ Scientific M970D8
Description
Performing a large number of biopsies means an increase in the number of containers handled, and consequently a technical overload of the transmission network, which occurs without any financial counterpart. A new approach had to be developed in order to increase productivity. The CoreDish is a multi-compartment container in the shape of a dish and half prefilled with 10% Neutral Buffered Formalin, for holding and transporting biopsies. Thanks to this innovative biopsy container, it is no longer necessary to use a multitude of individual containers, thereby reducing risk of confusion.
- Multi-compartment container is constructed of polystyrene
- Leakproof screw closure with O-ring ensures total protection of contents
- Writing area for patient information is provided on the label
- Each compartment is clearly identified to allow proper placement and visualization of the biopsy being inserted
- Dimensions: 15 H x 95 mm dia.
- For up to 5, 8 or 12 biopsies (depending on model)
- Compartments are numbered from 1 to 5, 1 to 8, or 1 to 12
- For breast biopsies
- Out of five compartments, four are labelled: LUQ, RUQ, LLQ, RLQ
Prostate
- A multi-compartment container for prostate biopsies
- Eight compartment; six are labeled: Base, Lateral Base, Medium, Lateral Medium, Apex, Lateral Apex
- 12 compartment; all are labeled: L Base, R Base, L Lateral Base, R Lateral Base, L Lateral Medium, L Medium, R Medium, R Lateral Medium, L Lateral Apex, L Apex, R Apex, R Lateral Apex
- For upper GI track biopsies
- Eight compartments; seven are labeled: Gastric Card, Gastric Body, GE Junction, Gastric ATR, Distal Oesophage, Pylorus, Duodeum
- For lower GI track biopsies
- Twelve compartments; ten are labeled: Proximal Traverse Colon, Hepatic Flexure Colon, Distal Transverse Colon, Ascending Colon, Splenic flexure Colon, Cecum, Decending Colon, Terminal Ileum, Rectum, Sigmoid Colon
Chemical Identifiers
| Polystyrene | |
| Leakproof Screw Cap with O-Ring | |
| 15 mm |
| Conforms to OSHA directives. | |
| 95 mm |
Specifications
| Polystyrene | |
| Leakproof Screw Cap with O-Ring | |
| 15 mm | |
| General Purpose |
| Conforms to OSHA directives. | |
| 95 mm | |
| 8 Compartments |