Learn More
Simport™ Scientific T100 BioTube™ Racks
The T100 BioTube™ Racks rack with the standard 96-well on-center spacing of tubes offers a color-coding system using colored interchangeable plastic grids.
Supplier:  Simport™ Scientific T1001Y
 View more versions of this product
Catalog No. 05-500-3
Only null left
Explore available promotions
Description
Description
  • Made of polypropylene
  • Free of RNase, DNase, Pyrogen, and DNA
  • Convenient carrying handles on both sides
  • Cover, grid plate, and base are keyed to prevent misalignment
  • Easy to read ID numbers and letters
  • Grid plate stands on 4 legs and can be placed on a lab counter or in a water bath
  • These racks and tubes are also ideal for storing, freezing, and transporting reagents and specimens
Specifications
Specifications
Polypropylene
Polypropylene
Rack with 96 plain individual tubes
Non-sterile
96-plain Tube Rack
Yellow
HTLV-III testing, bacterial and hybridoma cell uptake studies, cell harvesting
10/Cs.
Autoclavable
Product Suggestions
Product Suggestions