missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Simport™ Scientific T100 Tubes
T100 Tubes are made of autoclavable polypropylene and are available either individually or in strips of 8 or 12 detachable tubes. Ideal for HTLV-III testing, bacterial and hybridoma cell uptake studies, cell harvesting, pharmaceutical quality control, receptor binding assays, RIA, and EIA.
Supplier: Simport™ Scientific T10026
Description
- Tubes made of autoclavable polypropylene
- Free of RNase, DNase, Pyrogen, and DNA
- Compatible with ELISA, RIA, EIA procedures
- Available as an individual, 8-strip, and 12-strip configuration
- Gross volume of 1.2 mL
- Volume with cap is 1.1 mL
- Caps sold separately
Specifications
| 1.2 mL | |
| Polypropylene | |
| Strips of 12 tubes without writing surface | |
| Non-sterile | |
| 8.8 mm |
| 46 mm | |
| HTLV-III testing, bacterial and hybridoma cell uptake studies, cell harvesting | |
| 400/Cs. | |
| Autoclavable | |
| Culture Tube |