missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific Capinsert™ For Cryovial Tubes

Catalog No. 361015340
Change view
Click to view available options
Color:
Blue
Burnt Orange
Green
Lilac
Pink
Red
White
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
361015340 White
22038503 Burnt Orange
22038501 Yellow
22038504 Lilac
22038502 Pink
23045226 Blue
23045227 Red
23045228 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 361015340 Supplier Simport™ Scientific Supplier No. T3121

The Capinsert™ For Cryovial Tubes inserts fit precisely into the cap of the Cryovial™ for color identification.

  • Color coded inserts
  • Made of polypropylene
  • For Cryovial tubes

Specifications

Color White
Material Polypropylene
For Use With (Equipment) Cryovial™ Tubes