missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific Amplitube™ PCR Reaction Strips

Catalog No. 23045238
Change view
Click to view available options
Color:
Blue
Green
Natural
Red
Yellow
Closure Type:
Domed Cap
Flat Cap
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Type
23045238 Red Flat Cap
361024933 Natural Domed Cap
22039101 Natural Flat Cap
23045236 Blue Flat Cap
23045237 Green Flat Cap
23045239 Yellow Flat Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 23045238 Supplier Simport™ Scientific Supplier No. T3202R

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Made of polypropylene. Amplitube™ PCR Reaction Strips are manufactured under the most stringent conditions to attain the highest quality standards.

  • Available with either flat (needle pierceable) or dome-topped individually attached hinged caps
  • Hinged caps have integral seal that prevents contamination of lid surface
  • Ultra-thin sidewalls and bottoms for more uniform and efficient temperature transfers
  • Positive sealing fully protects contents from evaporation during thermal cycle
  • Strip fits standard well spacing and can also be used with 8-channel hand-held pipettors
  • Easily opened and closed with one hand
  • Autoclavable
  • Choice of colorless and four non-cytotoxic and non-metallic colors
  • Packed in tamper-proof resealable bags

Specifications

Color Red
Volume (Metric) 0.2 mL
Quantity 125 Pk.
For Use With (Equipment) Standard well spacing and can also be used with 8-channel hand-held pipettors
Sterility Non-sterile
Material Polypropylene
Closure Type Flat Cap
Autoclavable Autoclavable
Format Tube
Product Type 8-Strip PCR Reaction Tube