missing translation for 'onlineSavingsMsg'
Learn More

KNF Neuberger SIMDOS™ 10 Diaphragm Liquid Dosing Vacuum Pump

Liquid dosing/metering diaphragm pump. Offers wetted material combinations for optimal chemical resistance. Manual control of flow rate and dosing volume with LCD graphic display, also with optional analog and RS-232 control.

Supplier:  KNF Neuberger 167045160160

 View more versions of this product

Catalog No. 13880927


Only null left
Explore available promotions

Description

Description

  • Simple, intuitive setup and operation
  • Easy to calibrate in minutes
  • Runs dry without damage
  • Self-priming up to 9 ft. of water
  • Splash-proof IP65 housing
  • Analog and impulse logic control (RC) and with RS232 (RCP)
  • Small footprint conserves benchtop space
  • Adaptable to viscous fluid characteristics
Specifications

Specifications

RS232 Remote Control
Polypropylene Head
1 to 100 mL/min.
Diaphragm Vacuum Pump
Synthetic Organic Chemistry
100/240 V
150 x 93 x 144 mm
UFEM 1.10 KT.18RCP2
50/60 Hz
IP65
85 psig
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions