missing translation for 'onlineSavingsMsg'
Learn More

Silver wire, 2.0mm (0.08in) dia, hard, 99.9% (metals basis)

Catalog No. AA41457CD
Change view
Click to view available options
Quantity:
50 cm
1000 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41457CD 1000 cm
AA41457BW 50 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA41457CD Supplier Thermo Scientific Chemicals Supplier No. 041457CD

CAS: 7440-22-4 | Ag | 107.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Quantity 1000 cm
Odor Characteristic
Weight ≈32.94g/m
Form Wire
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Silver wire, 2.0mm (0.08 in.) dia., Hard
EINECSNumber 231-131-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only