missing translation for 'onlineSavingsMsg'
Learn More

Silver wire, 0.64mm (0.025in) dia, annealed, 99.9% (metals basis)

Form: Approx. 3.4 g/m; Temper: soft | CAS: 7440-22-4 | Ag | 107.87 g/mol

$120.04 - $1385.77

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA43325G2
View Documents
Thermo Scientific Chemicals
043325G2
2 m
Each for $120.04
Only null left
 
AA43325G6
View Documents
Thermo Scientific Chemicals
043325G6
10 m
Each for $349.53
Only null left
 
AA43325H2
View Documents
Thermo Scientific Chemicals
043325H2
50 m
Each for $1,385.77
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-22-4
107.87
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
CHEBI:9141
[Ag]

Specifications

Silver
Silver-colored wire
2 m
99.9% (Metals basis)
Ag
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
[Ag]
107.87
CHEBI:9141
7440-22-4
10.5g/cm3
Characteristic
Wire
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
23954
Silver, Annealed
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-131-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only