missing translation for 'onlineSavingsMsg'
Learn More

Silver wire, 0.127mm (0.005in) dia, annealed, 99.9% (metals basis)

Form: approx. 0.13g/m, Temper: soft | CAS: 7440-22-4 | Ag | 107.87 g/mol

Supplier:  Thermo Scientific Chemicals 000303G9

 View more versions of this product

Catalog No. AA00303G9


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Silver
7440-22-4
25 m
99.9%, metal basis
Ag
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
[Ag]
107.87
CHEBI:9141
Annealed
10.5g/cm3
Characteristic
Wire
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
23954

Chemical Identifiers

7440-22-4
107.87
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
CHEBI:9141
[Ag]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-131-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only