missing translation for 'onlineSavingsMsg'
Learn More

Silver wire, 0.05mm (0.002in) dia, annealed, 99.99% (metals basis)

Catalog No. p-7059355
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
2 x 50 m
5 m
50 m
Catalog No. Quantity
AA44461G5 5 m
AA44461H2 50 m
AA44461J4 2 x 50 m
3 options

Catalog No. AA44461G5

Supplier: Thermo Scientific Chemicals 044461G5

Only null left

CAS: 7440-22-4 | Ag | 107.87 g/mol

Silver wire finds application in electronics. It is also used in speaker systems for their magnet wire in order to get the best sound available from their speakers. Further, it is used in jewelry making.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Quantity 5 m
Odor Characteristic
Weight ≈0.02g/m
Form Wire
Solubility Information Insoluble in water.
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Silver wire, 0.05mm (0.002 in.) dia., Annealed
EINECSNumber 231-131-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only