missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver two part conductive adhesive, EXP. XX/XX Resin and Hardener
Description
A conductive epoxy which bonds well to metal, ceramics, plastics, glass and graphite
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Limited shelf life, 60 days after shipment
Specifications
Specifications
| Chemical Name or Material | Silver two part conductive adhesive |
| Name Note | EXP. x X/XX Resin and Hardener |
| Quantity | 1 g |
| MDL Number | MFCD00003397 |
RUO – Research Use Only