missing translation for 'onlineSavingsMsg'
Learn More

Silver tungsten oxide, 99% (metals basis)

Catalog No. AA3966118
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3966118 50 g
AA3966109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3966118 Supplier Thermo Scientific Chemicals Supplier No. 03966118

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13465-93-5 | Ag2O4W | 463.57 g/mol

Silver tungsten oxide is used in electronic and chemical industries. It is also used as a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13465-93-5
Molecular Formula Ag2O4W
Molecular Weight (g/mol) 463.57
MDL Number MFCD00014147
InChI Key JFUVYDSGHRJJDO-UHFFFAOYSA-N
Synonym Silver tungstate

Specifications

Color Yellow
Quantity 50 g
Odor Odorless
Sensitivity Light sensitive
Form Powder
Linear Formula Ag2WO4
Solubility Information Insoluble in water; Soluble in potassium cyanide, ammonium hydroxide and nitric acid
Formula Weight 463.58
Percent Purity 99%
Chemical Name or Material Silver tungsten oxide
EINECSNumber 236-708-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only