missing translation for 'onlineSavingsMsg'
Learn More

Silver Thinfoil, 0.005mm (0.0002in) thick, 99.97% (metals basis)

Catalog No. p-7051205
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40748FF 25 x 25 mm
AA40748FI 50 x 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA40748FF Supplier Thermo Scientific Chemicals Supplier No. 040748FF
Only null left

Microelectronics, contacts and coatings | CAS: 7440-22-4 | Ag | 107.87 g/mol

Microelectronics, contacts and coatings

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Quantity 25 x 25 mm
Odor Characteristic
Weight ≈0.03g/25x25mm
Form Foil
Percent Purity 99.97%
Assay (metals basis)
Chemical Name or Material Silver Thinfoil, 0.005mm (0.0002 in.) thick, Not certified pinhole free
EINECSNumber 231-131-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only