missing translation for 'onlineSavingsMsg'
Learn More

Silver Tetrafluoroborate 98.0+%, TCI America™

Catalog No. p-7135889
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04465G 5 g
S044625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S04465G Supplier TCI America Supplier No. S04465G
Only null left

Chemical Identifiers

CAS 14104-20-2
Molecular Formula AgBF4
Molecular Weight (g/mol) 194.67
MDL Number MFCD00003408
InChI Key CCAVYRRHZLAMDJ-UHFFFAOYSA-N
Synonym silver tetrafluoroborate, silver borofluoride, silver fluoborate, silver fluoroborate, silver i tetrafluoroborate, borate 1-, tetrafluoro-, silver 1+, silver tetrafluroborate, acmc-209clm
PubChem CID 159722
IUPAC Name silver(1+) tetrafluoroboranuide
SMILES [Ag+].F[B-](F)(F)F

Specifications

Melting Point 73°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
UN Number 1759
Formula Weight 194.67
Percent Purity ≥98.0% (T)
Chemical Name or Material Silver Tetrafluoroborate
EINECSNumber (1)-0001
RTECSNumber ED2875000
TSCA Yes