missing translation for 'onlineSavingsMsg'
Learn More

Silver Sulfate, puriss., Honeywell Fluka™

Catalog No. p-7071755
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001356 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6001356 Supplier Honeywell Chemicals Supplier No. 10229500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Puriss.,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10294-26-5
Molecular Formula Ag2H2O4S
Molecular Weight (g/mol) 313.81
MDL Number MFCD00003407
InChI Key YPNVIBVEFVRZPJ-UHFFFAOYSA-N
Synonym silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt
PubChem CID 159865
IUPAC Name disilver(1+) sulfuric acid
SMILES [Ag+].[Ag+].OS(O)(=O)=O

Specifications

Quantity 500 g
Linear Formula Ag2SO4
UN Number 3077
Formula Weight 311.8g/mol
Percent Purity ≥99%
Grade puriss.
Chemical Name or Material Silver Sulfate
Hazard Statement P280-P305 + P351 + P338 + P310
Precautionary Statement H318-H410
EINECSNumber 233-653-7