missing translation for 'onlineSavingsMsg'
Learn More

Silver sulfate, Premion™, 99.999% (metals basis)

Catalog No. p-4407828
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1211206 5 g
AA1211214 25 g
AA1211222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1211206 Supplier Thermo Scientific Chemicals Supplier No. 01211206
Only null left

CAS: 10294-26-5 | Ag2H2O4S | 313.81 g/mol

Silver(I) sulfate is employed in silver plating, and in electrolysis. It is a useful non-staining substitute to silver nitrate. Silver sulfate/iodine is an excellent iodination reagent for alkyl, alkoxybenzenes, aromatic amines and uridines. It is used in batteries; its pharmaceutical composition is used as an antimicrobial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10294-26-5
Molecular Formula Ag2H2O4S
Molecular Weight (g/mol) 313.81
MDL Number MFCD00003407
InChI Key YPNVIBVEFVRZPJ-UHFFFAOYSA-N
Synonym silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt
PubChem CID 159865
IUPAC Name disilver(1+) sulfuric acid
SMILES [Ag+].[Ag+].OS(O)(=O)=O

Specifications

Boiling Point 1085°C (decomposition)
Melting Point 657°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.999% (metals basis)
Solubility Information Partially soluble in water. Water solubility increases with temperature. Soluble in nitric acid,ammonia and conc. sulfuric acid.
Formula Weight 311.8
Odor Odorless
Sensitivity Light sensitive
Appearance White powder
Density 5.45 g/cm3 at 20°C
Chemical Name or Material Silver sulfate
Show More Show Less
Hazard Category H318
Precautionary Statement P280-P305+P351+P338-P310
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 233-653-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only