missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Sulfate (Powder/Certified ACS), Fisher Chemical™
Specifications
Specifications
| Melting Point | 652.2°C |
| Color | White |
| Physical Form | Solid |
| Quantity | 100 g |
| Assay Percent Range | ≥98 % |
| Formula Weight | 311.8 |
| Percent Purity | ≥98% |
| Grade | Certified ACS |
| Identification | Pass Test |
| Packaging | Glass Bottle |
| Show More |