missing translation for 'onlineSavingsMsg'
Learn More

Silver Sulfate, ACS Reagent, Reagents

Catalog No. p-200021010
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337772 100 g
01337771 25 g
01337770 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337772 Supplier Labchem Inc Supplier No. C1304000100C2
Only null left

Silver Sulfate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10294-26-5
Molecular Formula Ag2H2O4S
Molecular Weight (g/mol) 313.81
InChI Key YPNVIBVEFVRZPJ-UHFFFAOYSA-N
Synonym Sulfuric Acid silver salt
IUPAC Name disilver(1+) sulfuric acid
SMILES [Ag+].[Ag+].OS(O)(=O)=O

Specifications

Boiling Point 1085°C
Color Colorless/White
Physical Form Solid
Quantity 100 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 5.45
Chemical Name or Material Silver Sulfate, ACS Reagent