missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Sulfate, ACS Reagent, Reagents
Specifications
Specifications
| Boiling Point | 1085°C |
| Color | Colorless/White |
| Physical Form | Solid |
| Quantity | 25 g |
| Grade | ACS |
| Packaging | Glass Bottle |
| Specific Gravity | 5.45 |
| Chemical Name or Material | Silver Sulfate, ACS Reagent |