missing translation for 'onlineSavingsMsg'
Learn More

Silver sulfate, 99%, extra pure

Catalog No. p-3735351
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC209470100 10 g Glass bottle
AC209471000 100 g Glass bottle
AC209475000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC209470100 Supplier Thermo Scientific Chemicals Supplier No. 209470100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10294-26-5 | Ag2H2O4S | 313.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10294-26-5
Molecular Formula Ag2H2O4S
Molecular Weight (g/mol) 313.81
MDL Number MFCD00003407
InChI Key YPNVIBVEFVRZPJ-UHFFFAOYSA-N
Synonym silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt
PubChem CID 159865
IUPAC Name disilver(1+) sulfuric acid
SMILES [Ag+].[Ag+].OS(O)(=O)=O

Specifications

Boiling Point 1085.0°C
Melting Point 652.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 10 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula Ag2SO4
Fieser 01,1015; 03,254; 04,435
Merck Index 15, 8668
Solubility Information Solubility in water: 7.4g/L (20°C). Other solubilities: soluble in nh4oh, nitric acid and sulfuric acid, insoluble in alcohol
Formula Weight 311.79
Percent Purity 99%
Grade Extra Pure
Packaging Glass bottle
Density 1.84g/mL
Chemical Name or Material Silver sulfate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
EINECSNumber 233-653-7

RUO – Research Use Only