missing translation for 'onlineSavingsMsg'
Learn More

Silver slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, 99.99% (metals basis)

Catalog No. AA42921QK
Change view
Click to view available options
Quantity:
1 Slug
25 Slugs
5 Slugs
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42921QK 25 Slugs
AA42921QI 1 Slug
AA42921QJ 5 Slugs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA42921QK Supplier Thermo Scientific Chemicals Supplier No. 042921QK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-22-4 | Ag | 107.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Quantity 25 Slugs
Odor Characteristic
Weight ≈4.2g/slug
Form Slug
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Silver slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length
EINECSNumber 231-131-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only