missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, Premion™, 99.999% (metals basis)
Description
Silver slug is used in thermal evaporation and electrodes (Schottky diode and fast diode).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Silver |
| Appearance | Silver-colored slug |
| Quantity | 10 g |
| Odor | Characteristic |
| Form | Slug |
| Assay Percent Range | 99.999% (Metals basis) |
| Solubility Information | Insoluble in water. |
| Chemical Name or Material | Silver |
Safety and Handling
| EINECSNumber | 231-131-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only