Learn More
Silver slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, Premion™, 99.999% (metals basis)
CAS: 7440-22-4 | Ag | 107.87 g/mol
Supplier: Thermo Scientific Chemicals 04298414
Description
Silver slug is used in thermal evaporation and electrodes (Schottky diode and fast diode).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Silver slug | |
| 7440-22-4 | |
| Characteristic | |
| Slug | |
| Ag | |
| argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber | |
| BQCADISMDOOEFD-UHFFFAOYSA-N | |
| silver | |
| 23954 | |
| 99.999% |
| 3.175mm (0.125 in.) dia. x 3.175mm (0.125 in.) length | |
| 25 g | |
| ≈0.26g/slug | |
| (metals basis) | |
| MFCD00003397 | |
| Insoluble in water. | |
| [Ag] | |
| 107.87 | |
| CHEBI:9141 | |
| Premion |
Chemical Identifiers
| 7440-22-4 | |
| 107.87 | |
| BQCADISMDOOEFD-UHFFFAOYSA-N | |
| 23954 | |
| silver |
| Ag | |
| MFCD00003397 | |
| argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber | |
| CHEBI:9141 | |
| [Ag] |
Safety and Handling
EINECSNumber : 231-131-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only