missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Silver shot, 1-7mm (0.08-0.12in), Premion™, 99.999% (metals basis)

CAS: 7440-22-4 | Ag | 107.87 g/mol

Supplier:  Thermo Scientific™ 4739809

 View more versions of this product

Catalog No. AA4739809


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7440-22-4
Solid
MFCD00003397
[Ag]
107.87
10 g
Ag
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
99.999

Chemical Identifiers

7440-22-4
107.87
BQCADISMDOOEFD-UHFFFAOYSA-N
[Ag]
Ag
MFCD00003397
silver
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 30 Years

EINECSNumber : 231-131-3

SDS
Documents

Documents

Promotions

Promotions