missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Silver shot, 1-7mm (0.08-0.12in), Premion™, 99.999% (metals basis)

Catalog No. AA4739830
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4739830 250 g
AA4739809 10 g
AA4739818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4739830 Supplier Thermo Scientific™ Supplier No. 4739830

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-22-4 | Ag | 107.87 g/mol

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
IUPAC Name silver
SMILES [Ag]

Specifications

Quantity 250 g
Form Solid
Percent Purity 99.999
ShelfLife 30 Years
EINECSNumber 231-131-3