missing translation for 'onlineSavingsMsg'
Learn More

Silver rod, 12.5mm (0.492in) dia, Premion™, 99.9985% (metals basis)

Catalog No. AA11356BP
Change view
Click to view available options
Quantity:
2 cm
10 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA11356BP 2 cm
AA11356BS 10 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA11356BP Supplier Thermo Scientific Chemicals Supplier No. 011356BP

CAS: 7440-22-4 | Ag | 107.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Quantity 2 cm
Odor Characteristic
Weight ≈12.87g/cm
Form Rod
Percent Purity 99.9985%
Grade Premion
Assay (metals basis)
Chemical Name or Material Silver rod, 12.5mm (0.492 in.) dia.
EINECSNumber 231-131-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only