Learn More
Silver powder, spherical, APS 0.5-1 micron, 99.9% (metals basis)
Form: S.A. 1.5-2.5m2/g | CAS: 7440-22-4 | Ag | 107.87 g/mol
Supplier: Thermo Scientific Chemicals 04159718
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Silver | |
| 7440-22-4 | |
| 50 g | |
| 99.9% (Metals basis) | |
| Ag | |
| argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber | |
| [Ag] | |
| 107.87 | |
| CHEBI:9141 |
| Silver | |
| Silver-colored powder | |
| Characteristic | |
| Spherical Powder | |
| MFCD00003397 | |
| BQCADISMDOOEFD-UHFFFAOYSA-N | |
| silver | |
| 23954 |
Chemical Identifiers
| 7440-22-4 | |
| 107.87 | |
| BQCADISMDOOEFD-UHFFFAOYSA-N | |
| 23954 | |
| silver |
| Ag | |
| MFCD00003397 | |
| argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber | |
| CHEBI:9141 | |
| [Ag] |
Safety and Handling
H400-H410: Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
DOTInformation : DOT Class: 9, Packaging Group: III
EINECSNumber : 231-131-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only