missing translation for 'onlineSavingsMsg'
Learn More

Silver powder, spherical, APS 0.5-1 micron, 99.9% (metals basis)

Catalog No. AA4159709
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4159709 10 g
AA4159718 50 g
AA4159730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4159709 Supplier Thermo Scientific Chemicals Supplier No. 04159709

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Form: S.A. 1.5-2.5m2/g | CAS: 7440-22-4 | Ag | 107.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Color Silver
Appearance Silver-colored powder
Density 10.5g/cm3
Quantity 10 g
Odor Characteristic
Form Spherical Powder
Assay Percent Range 99.9% (Metals basis)
Chemical Name or Material Silver
Hazard Category H400-H410: Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 231-131-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only