missing translation for 'onlineSavingsMsg'
Learn More

Silver powder, spherical, -635 mesh, 99.9% (metals basis)

Catalog No. p-7051586
Change view
Click to view available options
Quantity:
10 g
250 g
Catalog No. Quantity
AA4160009 10 g
AA4160030 250 g
2 options

Catalog No. AA4160009

Supplier: Thermo Scientific Chemicals 04160009

Only null left

CAS: 7440-22-4 | Ag | 107.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
IUPAC Name silver
SMILES [Ag]

Specifications

Quantity 10 g
Mesh Size −635 Mesh
Odor Characteristic
Percent Purity 99.9%
Chemical Name or Material Silver powder
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 231-131-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only