missing translation for 'onlineSavingsMsg'
Learn More

Silver powder, spherical, -500 mesh, 99.9% (metals basis)

Catalog No. p-4824937
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4160209 10 g
AA4160218 50 g
AA4160230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4160209 Supplier Thermo Scientific Chemicals Supplier No. 04160209
Only null left

CAS: 7440-22-4 | Ag | 107.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Color Silver
Appearance Silver-colored powder
Quantity 10 g
Mesh Size -500 Mesh
Odor Characteristic
Form Spherical Powder
Assay Percent Range 99.9% (Metals basis)
Chemical Name or Material Silver
Hazard Category H400-H410; Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 231-131-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only