missing translation for 'onlineSavingsMsg'
Learn More

Silver powder, APS 0.7-1.3 micron, 99.9% (metals basis)

Catalog No. p-4782125
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Catalog No. Quantity
AA1445009 10 g
AA1445018 50 g
AA1445030 250 g
3 options

Catalog No. AA1445009

Supplier: Thermo Scientific Chemicals 01445009

Only null left

Form: S.A. 0.8-1.2m2/g | CAS: 7440-22-4 | Ag | 107.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Color Silver
Appearance Silver-colored powder
Density 10.5g/cm3
Quantity 10 g
Odor Characteristic
Form Powder
Assay Percent Range 99.9% (Metals basis)
Chemical Name or Material Silver
Hazard Category H400-H410; Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
Hazard Statement Use appropriate containment to avoid environmental contamination. This material and/or its container must be disposed of as hazardous waste.
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 231-131-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only