missing translation for 'onlineSavingsMsg'
Learn More

Silver powder, -120 mesh, Premion™, 99.995% (metals basis)

Catalog No. AA0078106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA0078106 5 g
AA0078114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA0078106 Supplier Thermo Scientific Chemicals Supplier No. 00078106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-22-4 | Ag | 107.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Physical Form Powder
Quantity 5 g
UN Number UN3077
Percent Purity 99.999%
Grade Premion
Odor Characteristic
Assay (metals basis)
Chemical Name or Material Silver powder
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 231-131-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only