missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver powder, -100 mesh, 99.95% (metals basis)
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Silver |
| Appearance | Silver-colored powder |
| Quantity | 100 g |
| Mesh Size | -100 Mesh |
| Odor | Characteristic |
| Form | Powder |
| Assay Percent Range | 99.95% (Metals basis) |
| Chemical Name or Material | Silver |
Safety and Handling
| Hazard Category | H400-H410; Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. |
| DOTInformation | DOT Class: 9, Packaging Group: III |
| EINECSNumber | 231-131-3 |
| RTECSNumber | VW3500000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only