missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver perchlorate monohydrate, 99.9% (metals basis)
Description
Silver perchlorate can be used in the manufacturing of explosives. It is generally used as a source of silver ions during silver nanoparticles synthesis. It is used as a catalyst in organic chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 485°C (decomposition) |
| Color | White |
| Physical Form | Crystalline |
| Quantity | 50 g |
| Odor | Odorless |
| Sensitivity | Hygroscopic; Light sensitive |
| Density | 2.800 g/mL |
| UN Number | UN1481 |
| Merck Index | 14,8523 |
| Solubility Information | Soluble in water and organic solvents. |
| Show More |
Safety and Handling
| Hazard Category | H272-H314 |
| Hazard Statement | GHS H Statement H272-H314-H318 May intensify fire oxidizer. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P210-P220-P221-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S. |
| EINECSNumber | 232-035-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only