missing translation for 'onlineSavingsMsg'
Learn More

Silver p-toluenesulfonate, 98+%

CAS: 16836-95-6 | C7H8AgO3S | 280.07 g/mol

Supplier:  Thermo Scientific Chemicals L0030714

 View more versions of this product

Catalog No. AAL0030714


Only null left
Explore available promotions

Description

Description

Silver p-toluenesulfonate is used to converts alkyl halides to tosylates, and benzyl selenyl chlorides into their corresponding selenyl tosylates. It used in promotion of the leaving ability of halogens. For use in the conversion of alkyl halides to alkyl tosylates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silver p-toluenesulfonate is used to converts alkyl halides to tosylates, and benzyl selenyl chlorides into their corresponding selenyl tosylates. It used in promotion of the leaving ability of halogens. For use in the conversion of alkyl halides to alkyl tosylates.

Solubility
Soluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Silver p-toluenesulfonate
25 g
MFCD00007511
silver p-toluenesulfonate, silver tosylate, silverp-toluenesulfonate, silver p-toluenesulphonate, silver 1+ tosylate, silver p-tosylate, benzenesulfonic acid, 4-methyl-, silver 1+ salt, p-toluenesulfonic acid silver salt, silver i 4-methylbenzenesulfonate, silver p-methylbenzenesulfonate
BRYKBDMLJJLFAB-UHFFFAOYSA-N
silver;4-methylbenzenesulfonate
85606
≥98%
Light sensitive
16836-95-6
C7H8AgO3S
UN3077
Soluble in water.
[Ag].CC1=CC=C(C=C1)S(O)(=O)=O
280.07
279.07
Odorless

Chemical Identifiers

16836-95-6
280.07
BRYKBDMLJJLFAB-UHFFFAOYSA-N
85606
[Ag].CC1=CC=C(C=C1)S(O)(=O)=O
C7H8AgO3S
MFCD00007511
silver p-toluenesulfonate, silver tosylate, silverp-toluenesulfonate, silver p-toluenesulphonate, silver 1+ tosylate, silver p-tosylate, benzenesulfonic acid, 4-methyl-, silver 1+ salt, p-toluenesulfonic acid silver salt, silver i 4-methylbenzenesulfonate, silver p-methylbenzenesulfonate
silver;4-methylbenzenesulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 240-859-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only