missing translation for 'onlineSavingsMsg'
Learn More

Silver p-toluenesulfonate, 98+%

Catalog No. AAL0030706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0030706 5 g
AAL0030714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0030706 Supplier Thermo Scientific Chemicals Supplier No. L0030706

CAS: 16836-95-6 | C7H8AgO3S | 280.07 g/mol

Silver p-toluenesulfonate is used to converts alkyl halides to tosylates, and benzyl selenyl chlorides into their corresponding selenyl tosylates. It used in promotion of the leaving ability of halogens. For use in the conversion of alkyl halides to alkyl tosylates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silver p-toluenesulfonate is used to converts alkyl halides to tosylates, and benzyl selenyl chlorides into their corresponding selenyl tosylates. It used in promotion of the leaving ability of halogens. For use in the conversion of alkyl halides to alkyl tosylates.

Solubility
Soluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 16836-95-6
Molecular Formula C7H8AgO3S
Molecular Weight (g/mol) 280.07
MDL Number MFCD00007511
InChI Key BRYKBDMLJJLFAB-UHFFFAOYSA-N
Synonym silver p-toluenesulfonate, silver tosylate, silverp-toluenesulfonate, silver p-toluenesulphonate, silver 1+ tosylate, silver p-tosylate, benzenesulfonic acid, 4-methyl-, silver 1+ salt, p-toluenesulfonic acid silver salt, silver i 4-methylbenzenesulfonate, silver p-methylbenzenesulfonate
PubChem CID 85606
IUPAC Name silver;4-methylbenzenesulfonate
SMILES [Ag].CC1=CC=C(C=C1)S(O)(=O)=O

Specifications

Quantity 5 g
UN Number UN3077
Solubility Information Soluble in water.
Formula Weight 279.07
Percent Purity ≥98%
Odor Odorless
Sensitivity Light sensitive
Chemical Name or Material Silver p-toluenesulfonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 240-859-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only