missing translation for 'onlineSavingsMsg'
Learn More

Silver oxide, 99%+, MilliporeSigma™

Catalog No. M1192080025
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1192080025 25 g Plastic bottle
M1192080100 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1192080025 Supplier Technidata Supplier No. 1.19208.0025

Chemical Identifiers

CAS 20667-12-3
Molecular Formula Ag2O
Molecular Weight (g/mol) 231.74
MDL Number MFCD00003404
InChI Key KHJDQHIZCZTCAE-UHFFFAOYSA-N
IUPAC Name (argentiooxy)silver
SMILES [Ag]O[Ag]

Specifications

Physical Form Powder
Quantity 25 g
Assay Percent Range 99%+
UN Number UN 3085
Formula Weight 231.74
Odor Odorless
Packaging Plastic bottle
Appearance Dark brown powder
Chemical Name or Material Silver oxide
Hazard Category H272: May intensify fire; oxidiser.
H314: Causes severe skin burns and eye damage.
EUH044: Risk of explosion if heated under confinement.
DOTInformation DOT Class: 5.1 (8), Packaging Group: II
EINECSNumber 243-957-1