missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate, 0.1 N standard solution

Catalog No. p-3735357
Change view
Click to view available options
Quantity:
1 L
5 L
Packaging:
HDPE Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC124250010 1 L HDPE Bottle
AC124250050 5 L HDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC124250010 Supplier Thermo Scientific Chemicals Supplier No. 124250010
Only null left

CAS: 7761-88-8 | AgNO3 | 169.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Volumetric standard solution

Specifications

Chemical Name or Material Silver nitrate
Name Note 0.1 N Standard Solution
Color Colorless
CAS 7761-88-8, 7732-18-5
Physical Form Liquid
Quantity 1 L
Molecular Formula AgNO3
Linear Formula AgNO3
MDL Number MFCD00003414
Fieser 01,1008; 02,366; 03,252; 04,429; 05,582; 07,321; 09,411; 10,350; 11,268; 12,257; 14,350; 15,39
Merck Index 15, 8657
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
Solubility Information Solubility in water: miscible.
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES [Ag+].[O-][N+]([O-])=O
IUPAC Name silver(1+) nitrate
Molecular Weight (g/mol) 169.87
PubChem CID 24470
ChEBI CHEBI:32130
Formula Weight 169.87
Grade Pure
Concentration or Composition (by Analyte or Components) 0.0998 to 0.1002N (20°C)
Packaging HDPE Bottle
Specific Gravity 1.015
Density 1.0150g/mL
Show More Show Less

RUO – Research Use Only