missing translation for 'onlineSavingsMsg'
Learn More

Silver nanopowder, APS 20-40nm, 99.9% (metals basis)

Catalog No. p-7059868
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4550906 5 g
AA4550914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4550906 Supplier Thermo Scientific Chemicals Supplier No. 04550906
Only null left

CAS: 7440-22-4 | Ag | 107.87 g/mol

Silver nanopowder is involved in the coating of silver and is used for medicinal purposes and in medical devices. Its larger surface area to volume ratio and greater activity of nano-silver are utilized in modern biochemical applications and drug delivery. It is also used in the fabrication of graphite epoxy nanocomposites and optical fiber sensors. Furthermore, it is involved in the formulation of water based ink.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Quantity 5 g
Odor Characteristic
Particle Size APS 20 to 40nm
Form Nanopowder
UN Number UN3077
Solubility Information Soluble in nitric acid.
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Silver nanopowder, No Oxide layer
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 232-033-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only