missing translation for 'onlineSavingsMsg'
Learn More

Silver N,N-Diethyldithiocarbamate 98.0+%, TCI America™

Catalog No. p-7126244
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D04905G 5 g
D049025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D04905G Supplier TCI America Supplier No. D04905G

Chemical Identifiers

CAS 1470-61-7
Molecular Formula C5H10AgNS2
Molecular Weight (g/mol) 256.13
MDL Number MFCD00004929
InChI Key NSVHDIYWJVLAGH-UHFFFAOYSA-M
Synonym silver diethyldithiocarbamate, unii-gvn001b86o, agddtc, silver n,n-diethylcarbamodithioate, diethyldithiocarbamic acid, silver salt, diethyl dithio carbamic acid silver salt, silver i diethyldithiocarbamate, silver, diethylcarbamodithioato-ks,ks', ditiocarb silver, silver n,n-diethyldithiocarbamate
PubChem CID 3034078
IUPAC Name silver;N,N-diethylcarbamodithioate
SMILES CCN(CC)C(=S)[S-].[Ag+]

Specifications

Color Yellow
Physical Form Crystalline Powder
Quantity 5 g
Linear Formula (C2H5)2NCS2Ag
Formula Weight 256.13
Percent Purity ≥98.0% (T)
Identification Passes Test
Chemical Name or Material Silver N,N-Diethyldithiocarbamate [for As analysis]