missing translation for 'onlineSavingsMsg'
Learn More

Silver Metal, Wool, Spectrum™ Chemical

Catalog No. p-7166266
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613138 1 g Amber Glass Bottle
18613140 5 g Amber Glass Bottle
18613139 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18613138 Supplier Spectrum Chemical Mfg Cor Supplier No. S10441GM
Only null left

Chemical Identifiers

CAS 7440-22-4
Molecular Weight (g/mol) 107.87

Specifications

CAS Max % 1
Quantity 1 g
Packaging Amber Glass Bottle
Form Fiber
Grade Ungraded
Chemical Name or Material Silver Metal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.