missing translation for 'onlineSavingsMsg'
Learn More

Silver iodate, 98%, Thermo Scientific Chemicals

Catalog No. p-4407812
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4010914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4010914 Supplier Thermo Scientific Chemicals Supplier No. 4010914

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7783-97-3
Molecular Formula AgIO3
Molecular Weight (g/mol) 282.77
MDL Number MFCD00014150
InChI Key YSVXTGDPTJIEIX-UHFFFAOYSA-M
Synonym silver iodate, unii-5p7ox4vt1f, silver 1+ ion iodate, 5p7ox4vt1f, silberjodat, silver 1+ iodate, acmc-20ajqa, iodic acid hio3 , silver 1+ salt, ag.io3, silver iodate 25g
PubChem CID 165642
IUPAC Name silver;iodate
SMILES [O-]I(=O)=O.[Ag+]

Specifications

Melting Point >200°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99%
Solubility Information Practically insoluble in water. Soluble in aqueous ammonia
Formula Weight 282.77
Odor Odorless
Sensitivity Light sensitive
Appearance White Powder
Density 5.53 g/cm3 at 20°C
Chemical Name or Material Silver iodate
Show More Show Less
Hazard Category H272-H315-H319-H335
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation DOT Class: 5.1, Packaging Group: II
EINECSNumber 232-039-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only