missing translation for 'onlineSavingsMsg'
Learn More

Silver(II) oxide, 99.9% (metals basis), Ag 86.6% min

CAS: 1301-96-8 | AgO | 123.867 g/mol

Supplier:  Thermo Scientific Chemicals 04018714

 View more versions of this product

Catalog No. AA4018714


Only null left
Explore available promotions

Description

Description

Silver(II) oxide is used to make silver oxide-zinc alkali batteries. It is also used as a reagent in laboratory reactions and forms transition metal-carbene complexes. It is also used as a catalyst to prepare silver metal. It acts as an effective carbon dioxide scrubber and used in nuclear submarines, the international space station and space shuttles. Further, it is used in the manufacture of filters for gas sensors and in some infection-resistant surgical fabric materials and fibrous textile articles. In addition to this, it is used in concrete and in some swimming pools to protect the water from undesired microbes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Silver(II) oxide
1301-96-8
Powder
(metals basis)
MFCD00044285
14,8522
Soluble in alkalis. Insoluble in water.
O=[Ag]
123.867
123.87
Ag 86.6% min.
Light and moisture sensitive
>100°C (decomposition)
Black to Gray
25 g
AgO
UN3085
silver ii oxide, silver oxide, silberperoxyd, silver oxide,ago, acmc-1aiyc, silver i,iii oxide, ksc203c6r, silver ii oxide 5g, silver ii oxide-ag
OTCVAHKKMMUFAY-UHFFFAOYSA-N
oxosilver
92152
99.9%
Odorless
7.44 g/mL

Chemical Identifiers

1301-96-8
123.867
OTCVAHKKMMUFAY-UHFFFAOYSA-N
92152
O=[Ag]
AgO
MFCD00044285
silver ii oxide, silver oxide, silberperoxyd, silver oxide,ago, acmc-1aiyc, silver i,iii oxide, ksc203c6r, silver ii oxide 5g, silver ii oxide-ag
oxosilver
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H314-H318
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P370+P380+P375-P372-P380-P501c

H272-H314-H318-H335

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, CORROSIVE, N.O.S.

EINECSNumber : 215-098-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only