missing translation for 'onlineSavingsMsg'
Learn More

Silver(II) oxide, 98%

Catalog No. p-7049783
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2269506 5 g
AA2269514 25 g
AA2269522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA2269506 Supplier Thermo Scientific Chemicals Supplier No. 02269506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Manufacturing of silver oxide-zinc alkali batteries | CAS: 1301-96-8 | AgO | 123.867 g/mol

Silver(II) oxide is used to make silver oxide-zinc alkali batteries. It is also used as a reagent in laboratory reactions and forms transition metal-carbene complexes. It is also used as a catalyst to prepare silver metal. It acts as an effective carbon dioxide scrubber and used in nuclear submarines, the international space station and space shuttles. Further, it is used in the manufacture of filters for gas sensors and in some infection-resistant surgical fabric materials and fibrous textile articles. In addition to this, it is used in concrete and in some swimming pools to protect the water from undesired microbes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1301-96-8
Molecular Formula AgO
Molecular Weight (g/mol) 123.867
MDL Number MFCD00044285
InChI Key OTCVAHKKMMUFAY-UHFFFAOYSA-N
Synonym silver ii oxide, silver oxide, silberperoxyd, silver oxide,ago, acmc-1aiyc, silver i,iii oxide, ksc203c6r, silver ii oxide 5g, silver ii oxide-ag
PubChem CID 92152
IUPAC Name oxosilver
SMILES O=[Ag]

Specifications

Melting Point >100°C (decomposition)
Color Black to Gray
Physical Form Powder
Quantity 5 g
UN Number UN3085
Merck Index 14,8522
Solubility Information Insoluble in water; Soluble in alkalies,ammonium hydroxide with decomposition and N2 formation.
Formula Weight 123.87
Percent Purity 98%
Odor Odorless
Sensitivity Light and moisture sensitive
Density 7.44 g/mL
Chemical Name or Material Silver(II) oxide
Show More Show Less
Hazard Category H272-H314-H318-H335
Hazard Statement GHS H Statement
H272-H314-H318
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P370+P380+P375-P372-P380-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, CORROSIVE, N.O.S.
EINECSNumber 215-098-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only