Learn More
Silver(II) fluoride, 98+%
Moisture and Light Sensitive | CAS: 7783-95-1 | AgF2
$87.90 - $1032.70
Chemical Identifiers
| CAS | 7783-95-1 |
|---|---|
| Molecular Formula | AgF2 |
| MDL Number | MFCD00003411 |
| Synonym | Silver difluoride |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA1161004
|
Thermo Scientific Chemicals
01161004 |
2 g |
Each for $87.90
|
|
|||||
|
AA1161009
|
Thermo Scientific Chemicals
01161009 |
10 g |
Each for $291.27
|
|
|||||
|
AA1161018
|
Thermo Scientific Chemicals
01161018 |
50 g |
Each for $1,032.70
|
|
|||||
Description
Silver(II) fluoride is used as a fluorinating and oxidizing agent. It is also used in the preparation of organic perfluorocompounds and in the fluorination of aromatic compounds. In addition, it is also used in pharmacy and electroplating industry. It serves as an intermediate in the catalysis of gaseous reactions with fluorine by silver. It is utilized in the oxidation of xenon to xenon difluoride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7783-95-1 | |
| AgF2 | |
| MFCD00003411 | |
| Silver difluoride |
Specifications
| 690°C | |
| 7783-95-1 | |
| 2 g | |
| AgF2 | |
| Silver difluoride | |
| 145.87 | |
| Brown crystals | |
| Silver(II) fluoride |
| Brown | |
| Crystalline | |
| 98+% | |
| MFCD00003411 | |
| Soluble in hexane,chloroform,chlorofluorocarbon and hydrogen fluoride. | |
| Light and moisture sensitive | |
| 4.6 |
Safety and Handling
P210-P220-P221-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H272-H302+H312+H332-H314-H335
DOTInformation : DOT Class: 8, Packaging Group: II
EINECSNumber : 232-037-5
RTECSNumber : VW4200000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only