missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver ICP Standard, 10,000 ppm Ag in 5% HNO3, Ricca Chemical
Supplier: Ricca Chemical Company PAG10KN100
Specifications
| Silver ICP Standard | |
| 7761-88-8,7697-37-2,7732-18-5 | |
| 10,000ppm Ag | |
| 1 mL = 10 mg Ag | |
| MFCD00003414 | |
| [Ag+].[O-][N+]([O-])=O | |
| 169.87 | |
| CHEBI:32130 |
| Amber Poly Bottle | |
| 90.0,6.75,1.42 | |
| 94.0,7.03,1.47 | |
| AgNO3 | |
| SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
| silver(1+) nitrate | |
| 24470 |