missing translation for 'onlineSavingsMsg'
Learn More

Silver(I) Trifluoromethanethiolate 95.0+%, TCI America™

Catalog No. S09775G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S09775G 5 g
S09771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S09775G Supplier TCI America Supplier No. S09775G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 811-68-7
Molecular Formula CHAgF3S
Molecular Weight (g/mol) 209.94
MDL Number MFCD25563234
InChI Key SVPQZGWGYCARGG-UHFFFAOYSA-N
Synonym Trifluoromethanethiol Silver(I) Salt
PubChem CID 24191737
IUPAC Name trifluoromethanethiol silver
SMILES [Ag].FC(F)(F)S

Specifications

Quantity 5 g
Formula Weight 208.93
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Silver(I) Trifluoromethanethiolate