missing translation for 'onlineSavingsMsg'
Learn More

Silver(I) oxide, 99.99% (metals basis)

Catalog No. aa4257714
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
25 g
4 x 25 g
5 g
Catalog No. Quantity
AA4257714 25 g
AA4257703 1 g
AA4257706 5 g
AA4257749 4 x 25 g
4 options

Catalog No. AA4257714

Supplier: Thermo Scientific Chemicals 04257714

Only null left

CAS: 20667-12-3 | Ag2O | 231.74 g/mol

Polishing glass, coloring glass yellow, catalyst, purifying drinking water, lab reagent, carbon dioxide scrubber, and chemical sensors. It is used in the preparation of other silver compounds, and silver-oxide batteries. In organic chemistry, silver oxide finds use as an oxidizing agent for aldehyes to produce carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20667-12-3
Molecular Formula Ag2O
Molecular Weight (g/mol) 231.74
MDL Number MFCD00003404
InChI Key KHJDQHIZCZTCAE-UHFFFAOYSA-N
IUPAC Name (argentiooxy)silver
SMILES [Ag]O[Ag]

Specifications

Melting Point 230°C (decomposition)
Color Brown
Physical Form Powder
Quantity 25 g
UN Number UN1479
Merck Index 14,8521
Solubility Information Soluble in dilute nitric acid and ammonia. Practically insoluble in water and alcohol
Formula Weight 231.74
Percent Purity 99.99%
Odor Odorless
Sensitivity Light sensitive
Density 7.2
Assay (metals basis)
Chemical Name or Material Silver(I) oxide
Show More Show Less
Hazard Category H271-H318
Hazard Statement GHS H Statement
H271-H318;
May cause fire or explosion; strong oxidizer.
Causes serious eye damage.
Precautionary Statement P210-P220-P221-P280-P283-P305+P351+P338-P306+P360-P310-P370+P378q-P371+P380+P375-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: I
EINECSNumber 243-957-1
RTECSNumber VW4900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only