missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver(I) oxide, 99.99% (metals basis)
Description
Polishing glass, coloring glass yellow, catalyst, purifying drinking water, lab reagent, carbon dioxide scrubber, and chemical sensors. It is used in the preparation of other silver compounds, and silver-oxide batteries. In organic chemistry, silver oxide finds use as an oxidizing agent for aldehyes to produce carboxylic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 230°C (decomposition) |
| Color | Brown |
| Physical Form | Powder |
| Quantity | 5 g |
| Assay Percent Range | 99.99% (metals basis) |
| Solubility Information | Soluble in dilute nitric acid and ammonia. Practically insoluble in water and alcohol |
| Formula Weight | 231.74 |
| Odor | Odorless |
| Sensitivity | Light sensitive |
| Appearance | Dark brown powder |
| Show More |
Safety and Handling
| Hazard Category | H271-H318 |
| Precautionary Statement | P210-P220-P221-P280-P283-P305+P351+P338-P306+P360-P310-P370+P378q-P371+P380+P375-P501c |
| DOTInformation | DOT Class: 5.1; Packaging Group: I |
| EINECSNumber | 243-957-1 |
| RTECSNumber | VW4900000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only